Autoclave Bag Red with Heat Indicator 250°F, Hardy Diagnostics
Catalog #: CA52428-868
Supplier: HARDY DIAGNOSTICS CA
Specifications
- Description:Autoclave bag
- Size:8×12"
- Cat. no.:CA52428-868
- Packaging:Bag
- Supplier No.:AB5819
Specifications
About this item
Autoclave bag is made of tough, thick polypropylene, double seamed bottom, solid red bag with black biohazard warnings in English and Spanish.
- Dimension: 8×12"
- Thickness: 2 mil
- Bag is rated to withstand temperatures up to 250 °F
Storage temperature: 15...30 °C. Heat indicator strip turns black when exposed to steam, OSHA approved labeling, plastic twist ties for closure furnished upon request.
Packaging: 400 per case
Customers Who Bought This Also Bought