Autoclave Bag Red with Heat Indicator 300°F, Hardy Diagnostics
Catalog #: CA52428-866
Supplier: HARDY DIAGNOSTICS CA
Specifications
- Description:Autoclave bag
- Size:19×24"
- Cat. no.:CA52428-866
- Packaging:Bag
- Supplier No.:AB5013
Specifications
About this item
Autoclave Bag is made of tough, thick polypropylene, double seamed bottom, solid red bag with black biohazard warnings in English and Spanish.
- Thickness 2 mil
- With heat indicator
- Bag is rated to withstand heat up to 300 °F
Storage temperature: 15...30 °C. Heat indicator strip turns black when exposed to steam, OSHA approved labeling, plastic twist ties for closure furnished upon request.
Packaging: 200 per case.
Customers Who Bought This Also Bought