ToxinSensor™ Chromogenic LAL Endotoxin Assay Kit, Genscript®
Catalog #: 10127-556
Supplier: Genscript
CAS Number:
Specifications
- Description:Chromogenic LAL Endotoxin Assay Kit
- Size:1 Kit (16 rxns)
- Cat. no.:10127-556
- Supplier No.:L00350C
Specifications
About this item
Genscript ToxinSensorTM Chromogenic LAL Endotoxin Assay Kit is designed to be a quantitative In Vitro end-point endotoxin test for human and animal parenteral drugs, biological products, and medical devices.
- Highly Sensitive detect endotoxin concentration in the range of 0.005 - 1 EU/mL
- Incubation period can be shortened to 14 minutes
- Color-stabilizer ensures accurate results
This method utilizes a modified Limulus Amebocyte Lysate and a synthetic color producing substrate to detect endotoxin chromogenically. In addition, any sample with color (e.g. cell bacterial culture medium, serum or blood) cannot be assayed by this kit.
Customers Who Bought This Also Bought