Order Entry
ContactUsLinkComponent
Flask volumetric
VWR® Volumetric Flask, Clear Glass, Class A, Serialized, with Penny Head Glass Stopper
Catalog #: 10124-382
Supplier:  VWR International
Flask volumetric
VWR® Volumetric Flask, Clear Glass, Class A, Serialized, with Penny Head Glass Stopper
Catalog #: 10124-382
Supplier:  VWR International

Specifications

  • Capacity:
    1 ml
  • Cat. no.:
    10124-382
  • Stopper No.:
    8
  • Stopper Type:
    Glass pennyhead
  • Certification:
    Individual
  • Tolerance:
    0.01 ml

Specifications

About this item

Made from borosilicate 3.3 expansion glass.

  • Borosilicate glass construction
  • Individually serialized
  • Glass penny head stopper

Flasks are serialized and supplied with glass penny head stopper.

Customers Who Bought This Also Bought