VWR® Wide Mouth Volumetric Flasks with PE Stoppers, Class A, Serialized
Catalog #: 10124-200
Supplier: VWR International
Specifications
- Capacity:100 ml
- Cat. no.:10124-200
- Stopper No.:16
- Stopper Type:Polyethylene
- Certification:Individual
Specifications
About this item
Made from borosilicate 3.3 expansion glass.
- Borosilicate glass construction
- Individually serialized
- Provided with polyethylene stopper
- Wide-mouthed design
Flasks are serialized and supplied with polyethylene stopper.
Ordering information: Flasks come with individual certificate of compliance.
Customers Who Bought This Also Bought