To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Chlorine Water
- Size:1 L
- Chemical/Physical Specifications:Saturated
- Cat. no.:470300-696
- Grade:Reagent
- Supplier No.:CC0270-1LR
- Disposal Method:This disposal method is published for your convenience. You MUST have checked with your federal, state, and local regulations before using this method and these methods are only applicable for small laboratory sized quantities.
Use a lab apron, chemical resistant gloves and goggles before following the below method to dispose of hazardous waste.
1. Open the bottle in a fume hood and leave overnight.
a. Be careful, as chlorine is a toxic gas. Make sure the fume hood is closed and running.
2. The chlorine in the solution will evaporate and then the remaining liquid can be poured down the drain. - Solution Preparation:There are three methods to prepare chlorine water for use in the classroom. All methods should take place in a fume hood. Method 1: Bubble chlorine gas from a gas cylinder into water. Method 2: Set up a gas generating apparatus with a funnel and delivery tube in a 2 hole rubber stopper. Add about 10 g of manganese dioxide (MnO2) to a flask. Slowly add about 40 ml of hydrochloric acid (HCl) and warm the flask with the rubber stopper inserted. The gas that is produced needs to be bubbled into a flask containing about 1 L of water. Method 3: Mix 100 ml of commercial bleach with 25 ml of 6 M hydrochloric acid. Dilute to a final volume of 1 L with water. Leave the container open until the bubbling stops. It may take days for the solution to stop bubbling. Once the solution stops bubbling, cap the bottle with the chlorine water in it.
Specifications
About this item
CAS Number: Mixture
Formula Weight: Mixture
Formula: Mixture
Hazard Info: Toxic, Corrosive
Density (g/mL): 1.02
Boiling Point (°C): Decomposes
Freezing Point (°C): 0
Solubility: Water
Synonyms: Chlorine Test Solution
Shelf Life (months): 36
Storage: White
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.