Order Entry
ContactUsLinkComponent
Vial
Cardboard Storage Boxes, Globe Scientific
Catalog #: 10118-764
Supplier:  Globe Scientific
Vial
Cardboard Storage Boxes, Globe Scientific
Catalog #: 10118-764
Supplier:  Globe Scientific

Specifications

  • Color:
    Red
  • Description:
    81-Place (9 x 9) Storage Boxes
  • Cat. no.:
    10118-764
  • For Use With:
    Up to 2" x 13 mm Tubes
  • Supplier No.:
    3091R

Specifications

About this item

These cardboard storage boxes are designed for the compact storage of sample tubes, cryogenic vials, and centrifuge tubes in a temperature range of –196 to 121°C.

  • Liquid-resistant coating
  • Designed for use in refrigerators and cryogenic freezer systems
  • Drainage holes eliminate excess liquid and reduce moisture build-up
  • Numbered grid on the inside bottom of each box aids in sample identification
Ordering information: Assorted colors are available for special order.
Delivery: Supplied complete with dividers.
Customers Who Bought This Also Bought