Order Entry
ContactUsLinkComponent
Biotunnel mice cardboard 100bx, 3 7/8Inch long x 2Inch inside diameter x 1/8Inch wall
Mouse and Rat Bio-Tunnels, Cardboard, Bio-Serv
Catalog #: CA89412-146
Supplier:  Bio-Serv
Biotunnel mice cardboard 100bx, 3 7/8Inch long x 2Inch inside diameter x 1/8Inch wall
Mouse and Rat Bio-Tunnels, Cardboard, Bio-Serv
Catalog #: CA89412-146
Supplier:  Bio-Serv

Specifications

  • Ø:
    5.1 cm (2")
  • Length:
    9.8 cm (3⅞")
  • Description:
    Bio-Tunnels for Mice
  • Cat. no.:
    CA89412-146
  • Packaging:
    100/box
  • Supplier No.:
    K3556

Specifications

About this item

Very economical sheltering, nesting and gnawing enrichment made of 100% recycled pulp paper

  • Helps reduce boredom and aggressive behavior and increases physical activity
  • Disposable
  • Not autoclavable; contains adhesives
  • Available sterile by gamma irradiation

Wall thickness: 0.32 cm (1/8")

Customers Who Bought This Also Bought