Order Entry
ContactUsLinkComponent
Wash bottle
VWR® Wash Bottles, Right-to-Know, Safety-Vented, with GHS Labeling
Catalog #: 10111-972
Supplier:  VWR International
Wash bottle
VWR® Wash Bottles, Right-to-Know, Safety-Vented, with GHS Labeling
Catalog #: 10111-972
Supplier:  VWR International

Specifications

  • Capacity:
    500 ml (16.9 oz.)
  • Description:
    70% Ethanol
  • Cat. no.:
    10111-972
  • Cap Color:
    Natural

Specifications

About this item

Safety-vented wash bottles provide compliance with the Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements - as pertains to 'Workplace Labels'.

  • Pre-printed with chemical name and formula
  • Unique valve design improves use and safety
  • Translucent LDPE construction with color-coded polypropylene closures

They are pre-printed with the chemical name and formula, GHS pictogram and signal word*, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.

A unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips, and allows dispensing in an upright or inverted position. The 53 mm (2") wide mouth makes refilling easy. Bottles are translucent LDPE (Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm and Toluene is red LDPE) with color-coded polypropylene closures to aid in quick and easy identification.

Note: *Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word. Methyl Ethyl Ketone and Toluene assemblies do not include vented valve.

Ordering information: Assortment orders include four bottles per bag, with one each of, Acetone, Isopropanol, Bleach, and Ethanol.
Customers Who Bought This Also Bought