Order Entry
ContactUsLinkComponent
Mounting Medium 4oz 6x case6
Mounting Medium, Epredia™
Catalog #: CA84000-112
Supplier:  Thermo Scientific
CAS Number:  
Mounting Medium 4oz 6x case6
Mounting Medium, Epredia™
Catalog #: CA84000-112
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Mounting Medium
  • Size:
    118.3 mL (4 oz.)
  • Cat. no.:
    CA84000-112
  • Supplier No.:
    4112

Specifications

About this item

Store slides for prolonged periods of time without fading of stains with Epredia™ mounting medium.

  • Rapid drying
  • Toluene-based
  • Optically clear

Store slides for prolonged periods of time without fading of stains with Epredia™ mounting medium.

This toluene-based rapid drying mounting medium is miscible with all clearing agents including xylene substitutes. It is optically clear and will not cause fading of stains, even after prolonged storage. This mounting medium is miscible with all clearing agents, including xylene substitutes.

Ordering information: The 118.3 ml (4 oz.) bottle (84000-112) is a squeeze bottle with a twist-open/close cap.
Customers Who Bought This Also Bought