To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Cubitainer®
- Description:Formalin, Neutral Buffered (NBF), 10% Solution
- Size:5 gal.
- Cat. no.:CA84000-024
- Application:Fix and Store Tissue Specimens
- Supplier No.:5705
Specifications
About this item
Use Epredia™ Histology Fixatives for both specialized and general techniques. Formulas developed for specific tissue types and applications.
- Achieve brilliant cytoplasmic staining and complete dehydration
- Achieve fixation and dehydration of specimens simultaneously with uniquely buffered alcoholic formalin fixatives
- Patented formulations minimize over-dehydration in smaller biopsies
- Product identification bands make the products easy to identify and distinguish from other laboratory chemicals
- Reagents are excellent for lipid extraction allowing for increased paraffin infiltration
- Solvents may be used for special stains
- Reduce spills with easy-to-pour high-quality dehydrants
Use Epredia™ Histology Fixatives for both specialized and general techniques. Formulas developed for specific tissue types and applications.
Enhance nuclear detail in immunohistochemistry techniques with buffered zinc formalin, obtain similar results to 10% neutral buffered formalin. Unbuffered zinc formalin acts similarly too but without mercury or buffering salts. Epredia 37% Formaldehyde can be used for both histological applications as well as for storage and preservation of tissue specimens.
Signature Series™ Pen-Fix™ is a uniquely buffered alcoholic formalin fixative. Excellent as a frozen section fixation that fixes and dehydrates tissue samples simultaneously. Performs well with fatty or thick tissue samples as the penetration rate is three times faster than 10% neutral buffered formalin.
Carson’s Modified Buffered Formalin is excellent as a general use fixative and may also be used as a fixative for electron microscopy techniques. Epredia Vytac™ is a simple, safe and economical method for disposal of both 10% buffered and unbuffered formalin; including zinc formalin. Vytac™ solution breaks down formalin into inert components neutralizing more than 99% of waste formalin collected.