Order Entry
ContactUsLinkComponent
HistoGel* Specimen Processing Gel vial, 10ml, PK12
HistoGel™ Specimen Processing Gel, Epredia
Catalog #: CA83009-992
Supplier:  Thermo Scientific
CAS Number:  
HistoGel* Specimen Processing Gel vial, 10ml, PK12
HistoGel™ Specimen Processing Gel, Epredia
Catalog #: CA83009-992
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Processing Gel Vial
  • Cat. no.:
    CA83009-992
  • Supplier No.:
    HG-4000-012

Specifications

About this item

Encapsulate and retain even small, friable and viscous specimens during histological processing with Epredia™ HistoGel™ Specimen Processing Gel.

  • Penetrates specimen without allowing specimen to escape from gel
  • Outperforms other agarose gel mixtures or thromboplastin at a fraction of the cost
  • Eliminates unwanted discoloration around specimens on slides
  • Remains inside theparaffin block during cutting and virtually unnoticeable during sectioning

Encapsulate and retain even small, friable and viscous specimens during histological processing with Epredia™ HistoGel™ Specimen Processing Gel.

This gel encapsulates and retains specimens during histological processing, regardless of the specimens’ small, friable, or viscous nature. Chemical agents penetrate the specimen without allowing it to escape from the gel. The gel does not retain histological stains, eliminating unwanted discoloration around specimens on slides. It is virtually unnoticeable during sectioning and will not pop out of the paraffin block during cutting.

Ordering information: Starter Kit includes twelve 10ml (0.34oz.) HistoGel™ vials, two WonderBlock cooling blocks, a dry bath incubator, and 500 disposable pipets.
Customers Who Bought This Also Bought