FEP Tubing, Ace Glass Incorporated
Catalog #: 10099-224
Supplier: Ace Glass
Small Business Enterprise
Specifications
- Length:10'
- Tubing I.D.:⁵⁄₁₆"
- Tubing O.D.:¼"
- Description:FEP Tubing
- Cat. no.:10099-224
- Supplier No.:12687-14
Specifications
About this item
FEP (Fluorinated Ethylene Propylene) tubing is ideal for flow monitoring applications do to its high clarity.
- Made of high molecular weight resin that has the best stress crack resistance
Meets FDA requirements for repeated food contact and is chemically inert to almost all industrial chemicals and solvents. Excellent UV light transmission due to its low refractive index. Inside surface is smooth and nonstick, resulting in a low coefficient of friction.
Customers Who Bought This Also Bought