Vial Caps with PTFE Faced Silicone Liner, Ace Glass Incorporated
Catalog #: 10098-800
Supplier: Ace Glass
Small Business Enterprise
Specifications
- Cap Size:20-400
- Cap Type:Closed Top
- Description:Vial Caps with PTFE Faced Silicone Liner
- Cat. no.:10098-800
- Supplier No.:5702-08
Specifications
About this item
White polypropylene vial screw cap features a PTFE faced silicone liner
Available with either closed or open top.
Customers Who Bought This Also Bought