To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen symbol:CYB5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10085-448
- Supplier No.:15469-1-AP
Specifications
About this item
CYB5B(Cytochrome b5 type B) is also named as CYB5M, OMB5 and belongs to the cytochrome b5 family. It is a 21 kDa membrane bound hemoprotein which function as an electron carrier for several membrane bound oxygenases. This protein is expressed on the plasma membrane of lymphoma cells but not normal lymphocytes, reactive lymphocytes or bone marrow precursor cells. The full length protein has a propeptide with 11 amino acids.
Western Blot: Mouse lung Tissue, 1:1000-1:10000; IHC: Human Testis Tissue, 1:20-1:200
Type: Primary
Antigen: CYB5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: