To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Caspase 9
- Clonality:Monoclonal
- Clone:1B7G2
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human,Mouse
- Antigen symbol:CASP9
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Antigen synonyms:EC 3.4.22.62|ICE-like apoptotic protease 6,ICE-LAP6,Apoptotic protease Mch-6,Apoptotic protease-activating factor 3|Apaf-3|CASP-9
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Protein A Purification
- Cat. no.:10084-098
- Supplier No.:66169-1-IG
Specifications
About this item
Caspase 9, apoptosis-related cysteine protease (CASP9,synonyms: MCH6, APAF3, APAF-3, ICE-LAP6, CASPASE-9c)is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce 2 subunits, large and small, that dimerize to form the active enzyme. Capase 9 is processed by caspase APAF1; this step is thought to be one of the earliest in the caspase activation cascade.
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human pancreas Tissue, 1:20-1:200
Type: Primary
Antigen: CASP9
Clonality: Monoclonal
Clone: 1B7G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse