Ultra Microtome Blades, Epredia™
Catalog #: CA89202-606
Supplier: Thermo Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:MX35 Ultra low-profile blades
- Cat. no.:CA89202-606
- Supplier No.:3053835
Specifications
About this item
Epredia disposable Ultra microtome blades use a proprietary steel hardening technology (Pink Technology) to achieve better cutting performance and longevity.
- Proprietary technology produces an edge stronger than traditional blades
- Special coating treatment provides durability and consistency from blade to blade
- High- and low-profile disposable blades can be used with any microtome or cryostat on the market
- Unique angle profile that facilitates faster ribboning
Customers Who Bought This Also Bought