Order Entry
ContactUsLinkComponent
Benzophenone, Lab Grade, (C6H5)2CO, 119-61-9, 25 g
Benzophenone
Catalog #: 470300-374
Supplier:  Ward's Science
CAS Number:  
Benzophenone, Lab Grade, (C6H5)2CO, 119-61-9, 25 g
Benzophenone
Catalog #: 470300-374
Supplier:  Ward's Science
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Benzophenone
  • Size:
    25 g
  • Chemical/Physical Specifications:
    Powder
  • Cat. no.:
    470300-374
  • Grade:
    Laboratory
  • Supplier No.:
    BB0108-25G

Specifications

About this item

CAS Number: 119-61-9
Formula Weight: 182.22
Formula: (C6H5)2CO
Boiling Point (°C): 581
Freezing Point (°C): 47
Shelf Life (months): 36
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Customers Who Bought This Also Bought