To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Sucrose
- Size:1 kg
- Chemical/Physical Specifications:Crystal
- Cat. no.:470302-810
- Grade:Laboratory
- Supplier No.:SS1020-1KG
Specifications
About this item
CAS Number: 57-50-1
Formula: C12H22O11
Density: 1.58g/mL
Freezing Point: 185.5°C
Solubility: Water
Synonyms: Cane Sugar, Beet Sugar
Shelf Life: 36 Months
CAS Number: 57-50-1
Density (g/mL): 1.58
Formula: C12H22O11
Formula Weight: 342.30
Freezing Point (°C): 185.5
Shelf Life (months): 36
Solubility: Water
Storage: Green
Please Note: This product is designed for educational and teaching laboratories and no certificate of analysis is available.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.