To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Gram's Iodine
- Size:500 mL
- Chemical/Physical Specifications:Aqueous, biological stain
- Cat. no.:470301-188
- Grade:Laboratory
- Supplier No.:IX0180-500ML
Specifications
About this item
CAS Number: Mixture
Density: 1.0 g/mL
Boiling Point: 100 °C
Freezing Point: 0 °C
Synonyms: Iodine-Potassium Iodide Solution, IKI, Iodine-Iodine, Iodine Solution, Iodine Lugol's Dilute, and Iodine 2%
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.