BulletKit® AGM® Astrocyte Growth Medium
Catalog #: CA10065-276
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:AGM® Astrocyte growth medium
- Cat. no.:CA10065-276
Specifications
About this item
Clonetics® AGM® Astrocyte Growth Medium has been developed to support the growth of primary astrocytes. Media products are provided as convenient BulletKits® which contain basal medium and necessary growth factors for cell culture.
Media are supplied in several formats:
Basal Medium - Contains no growth factors cytokines, or supplements
SingleQuots® - Growth factors, cytokines, and supplements (basel media not included)
BulletKits®- Basal media and SingleQuots® Kit packaged together
Delivery: Content:
1 × ABM® Basal Medium (CC-3187), 500 ml
1 × AGM® SingleQuots® Supplement Pack (CC-4123)
containing
1 × Bottle FBS, 15.0 ml
1 × Bottle L-Glutamine, 5.00 ml
1 × Vial GA-1000, 0.50 ml
1 × Vial Ascorbic Acid, 0.50 ml
1 × Vial HEGF, 0.50 ml
1 × Vial Insulin, 1.25 ml
1 × ABM® Basal Medium (CC-3187), 500 ml
1 × AGM® SingleQuots® Supplement Pack (CC-4123)
containing
1 × Bottle FBS, 15.0 ml
1 × Bottle L-Glutamine, 5.00 ml
1 × Vial GA-1000, 0.50 ml
1 × Vial Ascorbic Acid, 0.50 ml
1 × Vial HEGF, 0.50 ml
1 × Vial Insulin, 1.25 ml
Customers Who Bought This Also Bought