SingleQuots® AGM® Astrocyte Growth Medium Supplements and Growth Factors
Catalog #: CA10064-364
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:AGM® Astrocyte growth medium Singlequots® supplements and growth factors
- Cat. no.:CA10064-364
Specifications
About this item
Clonetics® AGM® Astrocyte Growth Medium has been developed to support the growth of primary human astrocytes. Clonetics® Media Products are provided as convenient BulletKits® which contain basal medium and necessary growth factors.
Basal medium and SingleQuots® Kit growth factors are available separately. Clonetics® Media Prodcuts are specially designed to support the growth of human and animal primary cells. Media systems have been specifically developed and optimized for various cell systems.
Media are supplied in several formats:
Basal Medium - contains no growth factors cytokines, or supplements
SingleQuots® - growth factors, cytokines, and supplements
BulletKits® - basal media and SingleQuots® Kit packaged together
Delivery: Content: Kit contains growth supplements for a complete growth medium, developed especially for astrocytes.
Customers Who Bought This Also Bought