Nucleofector® SG Cell Line 4D-X Kit S
Catalog #: CA10064-226
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:SG cell line 4D-Nucleofector® X kit S
- Cat. no.:CA10064-226
- No. of Reactions:32
- Supplier No.:V4XC-3032
Specifications
About this item
The SG Cell Line 4D-Nucleofector® X Kit S is one of our three 4D-Nucleofector® Kits that is suited for transfecting difficult-to-transfect cell lines in the 20 µL 16-well Nucleocuvette® Strip format when working with the 4D-Nucleofector® X Unit.
- Benefits:
- High transfection efficiencies for primary cells
- Cells maintain functionality post transfection
- Conditions are transferable between 4D-Nucleofector® System, 96-well Shuttle® System and 384-well Nucleofector® System
This kit has been determined to be optimal for transfecting following cell types:
Ba/F3
Colo-205, CHO-S
H4
NIH/3T3
Raji, Ramos
THP-1
WEHI
Applications:
Transfection of low cell numbers in the range of 5×10⁴ to 1×10⁶ cells.
Medium throughput applications.
Optimization of transfection conditions e.g. substrate titrations.
The kit is available in different variants for either 12 or 24 reactions.
Delivery: Content:
1×0.675 ml SF cell line Nucleofector® solution.
1×0.15 ml Supplement 1.
50 µg pmaxGFP® vector (1 μg/μl in 10 mM Tris pH 8.0).
2×16-well Nucleocuvette® strips (20 μl).
1×0.675 ml SF cell line Nucleofector® solution.
1×0.15 ml Supplement 1.
50 µg pmaxGFP® vector (1 μg/μl in 10 mM Tris pH 8.0).
2×16-well Nucleocuvette® strips (20 μl).
Customers Who Bought This Also Bought