4D-Nucleofector® X SE Cell Line Kit L
Catalog #: CA10064-154
Supplier: LONZA PHARMA - BIOSCIENCE CA
CAS Number:
Specifications
- Description:Optimal for transfecting cell types - 293-F (FreeStyle), 32D; A-20, A2058, A375, A431, A549, AGS, ARPE-19; BetaTC/6, BxPC-3; C6, CCRF-CEM, CHO-K1, CHO-S (Invitrogen), Cos-1; EL4; FDC-P1; H9c2 (2-1), HEK293, Hel9217, HepG2, High 5, HL-60, HMC-1-8, HT-1080, HT-29, Hut-78, HUV-EC-C; IMR-32; J774A.1; K-562; KG-1; L-428, LNCaP; MDA-MB-453, MDBK, Molt-4, MV-4-11; Nalm-6, NB-4, NCI-H1299 [H1299], Neuro-2a [N2a], NG108-15, NS0; P19, P815, PC-12, PC-3; Raw 264.7, RBL-1, RBL-2H3, Reh, Rin m5F, RPMI 8226; S2, S49, SaOS, SF9, SH-SY5Y, SK-BR-3, SK-N-AS, SK-N-MC, SK-N-SH, SK-OV-3, Sp2/0-Ag14, SW480; T2, T84, TF-1; U266B1, U-937; Vero
- Cat. no.:CA10064-154
- No. of Reactions:24
- Supplier No.:V4XC-2024
Specifications
About this item
The SE Cell Line 4D-Nucleofector® X Kit L is one of our three 4D-Nucleofector® Kits that is suited for transfecting difficult-to-transfect cell lines in the 100 µl Nucleocuvette® Vessel format when working with the 4D-Nucleofector® X Unit.
- High transfection efficiencies in difficult-to-transfect cell lines
- Cells maintain functionality post transfection
- Conditions are transferable between 4D-Nucleofector® System, 96-well Shuttle® System and 384-well Nucleofector® System
Applications: Transfection of cell numbers in the range of 2×10⁵ to 2×10⁷ cells, Low throughput applications.
The kit is available in different variants for either 12 or 24 reactions.
Delivery: Contents:
12 Reactions: 2 × 0.675 ml SE Cell Line Nucleofector® solution, 2 × 0.15 ml Supplement 1, 50 µg pmaxGFP® Vector (1 μg/μl in 10 mM Tris pH 8.0), 12 × 100 µl Nucleocuvette® Vessel, 12 × Single use pipettes.
24 Reactions: 2.25 ml SG Cell Line Nucleofector® solution, 0.5 ml Supplement 1, 50 µg pmaxGFP® Vector (1 μg/μl in 10 mM Tris pH 8.0), 24 × 100 µl Nucleocuvette® Vessel, 24 × Single use pipettes.
12 Reactions: 2 × 0.675 ml SE Cell Line Nucleofector® solution, 2 × 0.15 ml Supplement 1, 50 µg pmaxGFP® Vector (1 μg/μl in 10 mM Tris pH 8.0), 12 × 100 µl Nucleocuvette® Vessel, 12 × Single use pipettes.
24 Reactions: 2.25 ml SG Cell Line Nucleofector® solution, 0.5 ml Supplement 1, 50 µg pmaxGFP® Vector (1 μg/μl in 10 mM Tris pH 8.0), 24 × 100 µl Nucleocuvette® Vessel, 24 × Single use pipettes.
Customers Who Bought This Also Bought