Specifications
- Description:P100 + Nuisance organic vapors filter disk
- Environmentally Preferable:Safer Human and Environmental Health
- Mounting type:Keyless Bayonet
- Cat. no.:10065-358
- Supplier No.:7960
Specifications
About this item
The Moldex® 7000, 7800, and 9000 share a full line of cartridges and filters for the toughest of environments and most common applications.
- P100 Particulate filter disks allow for a wider field of vision due to a low profile design; they are encased in plastic for durability and rough handling
- P100 Particulate filter disks fit under welding helmets
- Use the 7940 P100 Particulate Filter Disk as a stand-alone or piggyback with any Moldex® gas/vapor cartridge using the 7920 piggyback adaptor
- Easy keyless bayonet mount eliminates mis-threading
- All product and packaging material is 100% PVC-free, for a greener alternative
The Moldex 7140 and 7640 come pre-assembled, but unlike competitive models, they can be easily disassembled. This feature enables the user to replace either the filter or the cartridge or both as they become used up, which helps to save money and reduce waste. With other brands, users are forced to discard the entire unit. COMPLIANCE JUST GOT A WHOLE LOT EASIER. Moldex® designed the 7000 and 7800 half masks and 9000 full face series to be the most user-friendly respirators on the market. These streamlined respirators feature fewer parts, minimal maintenance, and a wider field of vision. They are all ultra-lightweight, durable, and economical, so maximum comfort with extended use is built-in. The award-winning 7000/7800/9000 series add up to simple, comfortable, and economical respiratory protection that’s just plain easy to wear.