BASIK* 1.5 ml Screw top vial and cap eppendorf clear
Catalog #: 10063-394
Supplier: MicroSolv Technology Corp
BASIK* 1.5 ml Screw top vial and cap eppendorf clear
Catalog #: 10063-394
Supplier: MicroSolv Technology Corp
Customers Who Bought This Also Bought
MiniCollect* Tube K2E K2Edta Non sterile Lavender Cap 0.25/0.5 Ml PK500
Supplier: Greiner Bio-One
$656.98 to $2,627.82
Hydrogen peroxide 30%, ULTREX® II, Ultrapure for trace metal analysis, J.T.Baker®
Supplier: VWR International
$568.42
Potassium nitrate ≥99.0%, crystallised ACS, VWR Chemicals BDH®
Supplier: VWR International
$179.52 to $927.60
Copper, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, VWR Chemicals BDH®
Supplier: VWR International
$194.37
Hydrogen peroxide 29 - 32% (w/w) in aqueous solution stabilized ACS
Supplier: Thermo Scientific Chemicals
$126.01