BASIK* 1.5 ml Snaptop vial and cap slit eppendorf amber
Catalog #: 10063-388
Supplier: MicroSolv Technology Corp
BASIK* 1.5 ml Snaptop vial and cap slit eppendorf amber
Catalog #: 10063-388
Supplier: MicroSolv Technology Corp
Customers Who Bought This Also Bought
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
170,25 $ to 676,42 $
n-Heptane ≥99%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
359,51 $ to 1 103,97 $
Potassium Standard, 1 ml = 0.1 mg K, 100 ppm K, Ricca Chemical Company
Supplier: Ricca Chemical
118,85 $