Order Entry
ContactUsLinkComponent
Media Storage Glass Bottle 500ml, Graduated 80ml, 176x86mm, Chemical resistant, cs10
Duran Media Storage Glass Bottles, WHEATON®, DWK Life Sciences
Catalog #: 490013-168
Media Storage Glass Bottle 500ml, Graduated 80ml, 176x86mm, Chemical resistant, cs10
Duran Media Storage Glass Bottles, WHEATON®, DWK Life Sciences
Catalog #: 490013-168

Specifications

  • Cap Size:
    45 mm
  • Description:
    500 mL Bottles, Graduated 400 mL, O.D. 86 mm
  • Height:
    176 mm
  • Cat. no.:
    490013-168
  • Supplier No.:
    218014459

Specifications

About this item

Screw-cap glass media bottles are stable and chemically resistant.

  • Chemically resistant
  • Plastic pouring ring
  • Interchangeable caps and rings

They are complete with a plastic pouring ring which imparts drop-free working. Plastic caps and pouring rings are completely interchangeable since all bottles from 100 ml on have the same thread size. Bottle, pouring ring and cap can be sterilized (up to 140 ˚C).

Customers Who Bought This Also Bought