Order Entry
ContactUsLinkComponent
Membrane Protran Premium 0.45um NC
Amersham™ Protran® Premium Western Blotting Membranes, Nitrocellulose, Cytiva
Catalog #: CA10061-120
Supplier:  Cytiva
Membrane Protran Premium 0.45um NC
Amersham™ Protran® Premium Western Blotting Membranes, Nitrocellulose, Cytiva
Catalog #: CA10061-120
Supplier:  Cytiva

Specifications

  • Dimensions:
    300 mm x 4 m
  • Description:
    Nitrocellulose (NC) membrane, roll, pore size 0.45 µm
  • Supplier No.:
    10600003

Specifications

About this item

Amersham™ Protran® Premium nitrocellulose blotting membranes provide excellent sensitivity, resolution and low background in fluorescence Western blotting detection applications.

  • Available in popular pre-cut sizes
  • High binding capacity
  • Protran® Premium 0.2 NC offers low auto-fluorescence across a wide range of excitation/emission wavelengths, making the membrane suitable for fluorescence-based immunodetection and multiplexing
  • Protran® Premium 0.45 NC is an unsupported, 100% pure nitrocellulose membrane suitable for proteins of a wide range of molecular weights

Amersham Protran Premium offers excellent sensitivity, resolution, and low background, especially when used with the fluorescence Amersham ECL Plex Western Blotting Detection System.

Available with a choice of pore size: Protran® Premium 0.2 NC (with pore size 0,2 µm, binding capacity 173 to 203 µg IgG/cm²), or Protran® Premium 0.45 NC (with pore size 0,45 µm, binding capacity 162 to 180 µg IgG/cm²).

For convenience, Western blotting sandwiches (10 sandwiches/package) are also available. Each sandwich consists of pre-cut Amersham™ Protran® Premium NC membranes pre-assembled with 2×3 mm Chr filter papers.

Customers Who Bought This Also Bought