Order Entry
ContactUsLinkComponent
Tube Cap
Resprep® SPE Tube Parts & Accessories, Restek
Catalog #: 10059-448
Supplier:  Restek
Tube Cap
Resprep® SPE Tube Parts & Accessories, Restek
Catalog #: 10059-448
Supplier:  Restek

Specifications

  • Description:
    Tube caps, for 6 ml tubes
  • Material:
    PE
  • Cat. no.:
    10059-448
  • Supplier No.:
    26003

Specifications

About this item

Polyethylene and polypropylene parts and accessories are available for maintance of Resprep® SPE tubes.

  • For 20 ml packed tubes
  • Containment volume compatibility varies between part models

Choose from a variety of parts and accessories including frits, tube caps, empty tubes, female luer end caps, and tube connectors.

Customers Who Bought This Also Bought