GC Snap Ring Cap With Septum, 11 mm, Restek
Catalog #: 10058-678
Supplier: Restek
Specifications
- Septum:PTFE/Silicone
- Color:Clear
- Description:Snap Ring Cap w/Septum
- Material:PP
- Supplier No.:21734
- Orifice:11 mm
Specifications
About this item
Restek snap ring caps are an extremely sturdy, affordable option when crimp-top caps are not needed.
- Economical
- Sturdy polypropylene construction
Use these snap caps when you crimping is unnecessary. Snap caps are available with either liners or septa, and they seal an 11-millimeter opening.
Customers Who Bought This Also Bought