Order Entry
ContactUsLinkComponent
Seal
Aluminum Crimp Seals with Septa, 11 mm, Restek
Catalog #: 10058-634
Supplier:  Restek
Seal
Aluminum Crimp Seals with Septa, 11 mm, Restek
Catalog #: 10058-634
Supplier:  Restek

Specifications

  • Color:
    Silver
  • Description:
    Seals w/Septa
  • Material:
    Aluminum
  • Cat. no.:
    10058-634
  • Supplier No.:
    21174
  • Orifice:
    11 mm

Specifications

About this item

Aluminum seals provide a sturdy cap to crimp-top vials that, when turned, allow for easy removal of the septum.

  • Crimp-top vial compatibility
  • Economical
  • Silver aluminum

Aluminum seals are the preferred choice for use with manual crimping tools, and for autosamplers that do not employ magnetic pick-up transport.

11 mm aluminum seals are available with septa pre-inserted for convenience and to reduce contamination due to handling.

Ordering information: Choose preslit caps (available for some vials) to reduce the risk of needle bending, release vacuum from high-volume injections, and improve injection reproducibility when greater than 20% of vial volume is withdrawn.
Customers Who Bought This Also Bought