Finger-Tight Capillary Column Nuts, Restek
Catalog #: 10058-592
Supplier: Restek
Specifications
- Description:Finger-Tight Capillary Column Nuts
- Cat. no.:10058-592
- For Use With:for Agilent GCs
- Supplier No.:21040
Specifications
About this item
Finger-tight capillary column nuts are an excellent alternative to conventional column nuts.
- Available for standard (1/16") and compact (Agilent-style) ferrules
- Made from high-quality stainless steel
They can be installed by hand without a wrench, and they cool down much faster than conventional fittings, making column and inlet maintenance easier. These stainless steel fittings work best with graphite-type ferrules and help prevent over-tightening, which can cause graphite material to exude into the inlet.
Customers Who Bought This Also Bought