Order Entry
ContactUsLinkComponent
Mag-Bind* Blood DNA HDQ 96 kit
Mag-Bind® Blood DNA HDQ 96 Kit, Omega Bio-tek
Catalog #: CA89498-638
Supplier:  Omega Bio-Tek
CAS Number:  
Mag-Bind® Blood DNA HDQ 96 Kit, Omega Bio-tek
Catalog #: CA89498-638
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Blood
  • Target molecule:
    DNA
  • Description:
    Mag-Bind® Blood DNA HDQ 96 Kit
  • Size:
    1 x 96 Well Plate
  • Description:
    Mag-Bind® Blood DNA HDQ 96 Kit
  • Cat. no.:
    CA89498-638
  • Supplier No.:
    M6399-00

Specifications

About this item

The Mag-Bind® Blood DNA HDQ 96 Kit is designed for rapid and reliable isolation of high-quality genomic DNA from 100-200 μL blood samples

All heating steps that limit robotic applications have been removed to allow for faster processing. Mag-Bind® Particles HDQ provide quick magnetic response time reducing overall processing time. This system combines the reversible nucleic acid-binding properties of Mag-Bind® paramagnetic particles with the time-proven efficiency of Omega Bio-tek’s blood DNA isolation system to provide a fast and convenient method to isolated DNA from fresh or frozen blood. Utilizing paramagnetic particles provides high-quality DNA that is suitable for direct use in most downstream applications, such as amplification and enzymatic reactions.

Customers Who Bought This Also Bought