Order Entry
ContactUsLinkComponent
Mounting medium M800 aqua 20ml
Aqua-Mount® Mounting Medium, Thermo Scientific
Catalog #: CA41799-008
Supplier:  Thermo Scientific
CAS Number:  
Mounting medium M800 aqua 20ml
Aqua-Mount® Mounting Medium, Thermo Scientific
Catalog #: CA41799-008
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Aqua-Mount® Mounting Medium
  • Size:
    20 mL
  • Cat. no.:
    CA41799-008
  • Supplier No.:
    13800

Specifications

About this item

All Lerner reagents are manufactured using strictly-controlled processing procedures to ensure batch-to-batch consistency

  • Non-Fluorescing, Aqueous Formula for Immunostaining

All Lerner reagents are manufactured using strictly-controlled processing procedures to ensure batch-to-batch consistency

ISO-9000 certified quality procedures include verification of product quality through actual tissue testing. Dyes used are all certified by the Biological Stain Commission.

Customers Who Bought This Also Bought