ContactUsLinkComponent
Microtome Blade
Epredia™ Premier Microtome Blades
Catalog #: CA14011-013
Supplier:  Thermo Scientific
Microtome Blade
Epredia™ Premier Microtome Blades
Catalog #: CA14011-013
Supplier:  Thermo Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Premier Plus microtome blades
  • Supplier No.:
    3052835
Specifications
About this item
Disposable blades have a 34° facet angle and feature a superior edge profile in which the shape of the blade edge is optimized to provide excellent sectioning performance, easier ribboning, and improved sharpness.
Premier low profile blades for routine application with higher proportion of harder tissues.
Premier Plus low profile blades for sectioning hard tissue and high-quality thin sections.
Customers Who Bought This Also Bought