To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:5 ml (⅙ oz.)
- Membrane Material:Omega
- Color:Yellow
- Operating temperature range:0...40 °C (32...104 °F)
- Overall length:12.0 cm (142/3")
- Effective filtration area:3.3 cm2
- Max. centrifugal force:7500 g (ultrafiltion), 14000 g (microfiltration)
- MWCO:1K
- Supplier No.:MCP001C41
Specifications
About this item
Precise, quick recovery of microliter volumes of concentrate from starting volumes up to 5.0 ml.
- Available with low protein-binding OmegaTM and Supor® (PES) membranes
- Omega modified PES membranes come in a wide range of MWCOs, color-coded for easy identification
- With Omega membranes you can Achieve 50X concentration and >90% recovery in just minutes
- Supor (PES) membranes come in 0.45 and 0.2 µm pore sizes for microfiltration applications
- Constructed of low-binding polypropylene
- Ultrasonically welded seals prevent bypass or seal failure
For smaller samples use Pall Nanosep ®centrifuge filters. For larger samples use Pall Macrosep® or Jumbosep™ centrifugal filter.
Microsep® Centrifugal devices with Omega (modified polyethersulfone) membrane for ultrafiltration are ideal for concentration of dilute protein samples, buffer provides an efficient way to remove or exchange salts, as well as fractionating molecules of different molecular weights. Supor (PES) microfiltration devices are ideal for clarifying samples with gross particulate, as well as removing proteins and particulate from samples for HPLC analysis of drugs, amino acids, and antibodies.
A built-in deadstop feature prevents samples from spinning to dryness and the unique paddle shape design prevents the creation of a density gradient, preventing proteins from aggregating. Filters are color coded and laser etched for easy identification. The Microsep sample reservoir and filtrate receiver are constructed of polypropylene to reduce non-specific binding of biomolecules.