Open Top Screw Caps, Thermo Scientific
Catalog #: 10056-148
Supplier: Thermo Scientific
Specifications
- Cap Size:15 mm-425
- Description:Open-Top Screw Caps
- Cat. no.:10056-148
- Supplier No.:TS-13216
Specifications
About this item
Provide inert, air-tight seal and direct puncture-access to sample with a syringe needle.
Customers Who Bought This Also Bought