ContactUsLinkComponent
Repeater M4 Starter Kit
Eppendorf® Repeater® M4 Starter Kit
Catalog #: CA10054-526
Supplier:  Eppendorf
Repeater M4 Starter Kit
Eppendorf® Repeater® M4 Starter Kit
Catalog #: CA10054-526
Supplier:  Eppendorf
Specifications
  • Volume:
    1 µl - 10 ml
  • Description:
    Repeater® M4 Starter Kit
  • Supplier No.:
    4982000322
Specifications
About this item
Introductory set includes a sampling of volumes.
  • Easy to dispense repeatable volumes in assays
  • Multiple volumes available in package
  • Ergonomically designed
The Eppendorf® Repeater® M4 offers a variety of unique features. A step counter displays the number of executed dispensing steps. An ergonomically shaped operating lever dispenses the liquid and ejects the Combitip® advanced. An egonomically shaped filling lever is included for filling and emplying the Combitip® advanced. The volume selection dial offers twenty different volume settings for every Combitip® size, and is easy to reach and adjust using only the thumb. An integrated hand rest supports a relaxed hand position for increased ease-of-use. Units also feature a motion sensor, allowing the Repeater® M4 to switch off when not in use, and back on again with minimal movements to save power and time. Additionally, a Combitip® sensor is included to provide automatic Combitip® recognition and volume display.
Ordering information: Starter Kit includes Combitip® Rack, Combitip® assortment pack 1 µl to 10 ml dispensing volume (for use with Combitip® advanced 0.1 to 50 ml).
Customers Who Bought This Also Bought