Cell Strainers
Catalog #: 10054-458
Supplier: Corning
Specifications
- Pore size:100 µm
- Color:Yellow
- Supplier No.:431752
Specifications
About this item
Corning® cell strainers are sterile, easy to use devices for rapidly isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Fits 50 ml Falcon® conical tubes.
- Strong nylon mesh for optimal performance in a variety of applications
- Extended lip on strainer enables aseptic handling with forceps
- Moulded color-coded polypropylene frame with tab enables easy handling and identification
- Sterilised by gamma irradiation, non cytotoxic
- Conveniently accessible in individual packaging
Use these versatile cell strainers to prepare samples for flow cytometry, single cell suspensions of blood cells, specimens for primary cell cultures and immunogens, as well as primary cells from tissues.
Customers Who Bought This Also Bought