To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- pH:7.416
- Description:Traceable® CRM pH Buffer
- Size:500 mL
- Supplier No.:4286
Specifications
About this item
Traceable® pH Standards, Certified Reference Materials, are 100% compatible with all instruments and probes
Certified Reference Materials meet Federal/State/local agencies’ strictest mandates and deliver exact calibration results for any pH meter. Select values are manufactured to IUPAC (International Union of Pure and Applied Chemistry) formulation. All Certified Reference Materials feature resistance to temperature changes and long-term buffer stability.
ISO 17025 (Calibration Laboratory) and ISO Guide 34 (Certified Reference Material Producer) provide highest achievable levels of product production, documentation, and accuracy. Additional accreditations include ISO 31 (content) and ISO 35 (statistical analysis). ISO 9001 ensures that world-class product quality standards are always met.
To assure accuracy an individually serial-numbered Traceable® Certificate is supplied to indicate traceability to standards provided by NIST (National Institute of Standards and Technology) and/or a National Standards Laboratory. Each 100 ml container is supplied with an individual temperature compensation chart, traceability information, and Traceable® Certificate.
Accuracy for the Traceable® Buffers at 25°C is ±0.010 pH.