Specifications
- Skirt:Fully skirted
- Well Color:Clear
- Well Volume:150 µl
- Description:25 plates (5 bags × 5 plates)
- Environmentally Preferable:Low Manufacturing Impact
- Frame Color:Clear
- No. of wells:96
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Supplier No.:0030129512
Specifications
About this item
Eppendorf twin.tec® PCR plates LoBind, available in 96 and 384-well formats, maximize nucleic acid recovery and are certified as 'PCR clean' for consistently reliable results.
- LoBind material guarantees maximum sample recovery for improved assay results
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
- Exceptionally solid and torque-resistant polycarbonate frame
- OptiTrack® matrix for faster sample identification and fewer pipetting errors
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Available in 96- and 384-well formats, twin.tec® PCR plates LoBind® improve recovery of nucleic acids by reducing their adsorption to the wall of the wells. A combination of special manufacturing technologies and selected polypropylene batches ensures that nearly 100 % of DNA and RNA molecules are recovered without the use of surface coatings, thereby eliminating the risk of sample contamination.
Offering maximum confidence in results, all Eppendorf twin.tec PCR plates LoBind are'‘PCR clean'. This means the plates are batch-tested and certified by an independent laboratory to be free of a range of common PCR contaminants, including DNA, DNAse, RNAse, and PCR inhibitors.
Autoclavable: 121 °C, 20 min.
Centrifugation stability: 6000×g
Purity grade: PCR clean (nonsterile)