Charging Stands for Picus and eLINE® Pipettes, Sartorius
Catalog #: CA89171-670
Supplier: Sartorius
Specifications
- Description:Four-Position Charging Carousel
- Cat. no.:CA89171-670
- Supplier No.:730991
Specifications
About this item
1-place stand and 4-place carousel for charging and storing Picus or eLINE® pipettes.
The compact design of the 4-pipette charging carousel is ideal for saving bench space in the laboratory. The 4-place rotating head provides easy access to the desired unit.
Customers Who Bought This Also Bought