Benzyl chloride for synthesis, Sigma-Aldrich®
Catalog #: CA8.01809.0100
Supplier: MilliporeSigma
CAS Number: 100-44-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:100 mL
- Cat. no.:CA8.01809.0100
- Supplier No.:8.01809.0100
Specifications
Specification Test Results
- Solubility:460 mg/l (30 °C)
- Melting Point:-41.2 °C
- Molar Mass:126.58 g/mol
- Boiling Point:179.3 °C (1013 hPa)
- Vapor Pressure:1.2 hPa (20 °C)
- Flash Point:67 °C
- Refractive Index:1.5415
- Explosion Limit:1.1 - 14 %(V)
- Density:1.10 g/cm3 (20 °C)
- pH:(H2O) not applicable
- Ignition Point:585 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
- Sigma-Aldrich®, Benzyl chloride for synthesis:
Specification Test Results
About this item
Synonyms: (Chloromethyl)benzene,Chlorophenylmethane
Boiling Pt:179.3 °C (1013 hPa)
Melting Pt:–41.2 °C
Density:1.103 g/cm³ (20 °C)
ADR:6.1 (8),II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ200
Flash Pt:52 °C
:Boiling Pt:
:Boiling Pt: 179.3 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –41.2 °C
:Flash Pt:
:Flash Pt: 52 °C
:M.W. 126.59 g/mol
Merck Index:13,01131
EINECS:202-853-6
:179.3
Storage Temperature:Ambient
UN:1738
:6.1 (8)
:II
:52
:GHS06
:GHS05
:GHS08
:MQ200
Customers Who Bought This Also Bought