Order Entry
ContactUsLinkComponent
Non-filtered tip
Biohit Family Extended Length Pipette Tips, Sartorius
Catalog #: CA10052-302
Supplier:  Sartorius
Biohit Family Extended Length Pipette Tips, Sartorius
Catalog #: CA10052-302
Supplier:  Sartorius

Specifications

  • Volume:
    100 - 5000 µL
  • Packed:
    Bulk
  • Cat. no.:
    CA10052-302
  • Supplier No.:
    780308

Specifications

About this item

The non-filtered Optifit tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision.

  • Tips work with most brands of pipettors
  • Autoclavable at 121 °C (250 °F) for 20 minutes
  • Single-handed opening and closing of flip-top racks
  • E-beam presterilized for sterile work

Use to reach solutions in flasks, deep well plates, tubes, and other difficult vessels

These pipette tips guarantee accurate results when used with Biohit® pipettors.

The tips work with most popular brands of single and multichannel pipettors. They also fit most research-grade pipettors.

Customers Who Bought This Also Bought