ContactUsLinkComponent
Nitrate Test
Nitrate Test Method: Reflectometric With Test Strips 5 - 225 mg/L NO₃⁻ Reflectoquant®, MilliporeSigma
Catalog #: CA1.16971.0001
Supplier:  MilliporeSigma
CAS Number:  
Nitrate Test Method: Reflectometric With Test Strips 5 - 225 mg/L NO₃⁻ Reflectoquant®, MilliporeSigma
Catalog #: CA1.16971.0001
Supplier:  MilliporeSigma
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Chemical method:
    Griess' reaction
  • Description:
    Test with Strips
  • Cat. no.:
    CA1.16971.0001
  • Range:
    5 - 225 mg/l NO₃
  • Supplier No.:
    1.16971.0001
  • No. of Tests:
    50 strips
Specifications
About this item
The rapid determination of the nitrate content is of particular interest with fruit and vegetables to help decide whether a given delivery batch can be released for use in a specific product (e.g baby foods).
MilliporeSigma # 1.16971.0001 replaces and is identical to MilliporeSigma # 16971-1
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
Customers Who Bought This Also Bought