LiChroCART® Stainless Steel Superspher® Cartridges, MilliporeSigma
Catalog #: CA48219-610
Supplier: MilliporeSigma
Specifications
- Column Format:Cartridge
- I.D.:4.00 mm
- Length:75 mm
- Particle size:4 µm
- Pore size:100 Å
- Description:Superspher® 100 RP-18 LiChroCART® HPLC cartridge
- Cat. no.:CA48219-610
- Packing Material:Silica
- Phase:RP-18
- Includes:3 columns
- Supplier No.:1.50980.0001
Specifications
About this item
These cartridges are ideal for highly efficient HPLC analysis of complex mixtures where high peak capacity is required, because the guaranteed number of theoretical plates for Superspher® cartridges is <100.000 N/m.
These LiChroCART® cartridges are compatible with manu-CART® '4' parts, which facilitates faster and more flexible method adaptation to smaller bore columns. Endcapped cartridges have a bonded phase that is reacted with trimethylchlorosilane to cover silanols not previously reacted with the initial bonding reagent.
This high performance spherical silica carrier yields an excellent pressure/separation performance ratio according to theoretical calculation and practical experience.
Ordering information: All LiChroCART® cartridges with lengths of 75, 125, or 250 mm require manu-CART® end fittings (14229-736).
Customers Who Bought This Also Bought