VWR® Vacuum Filtration Systems, Standard Line
Catalog #: 10040-474
Supplier: VWR International
Specifications
- Capacity:1000 ml
- Pore size:0.45 µm
- Description:Filtration Cup
- Membrane Diameter:91 mm
Specifications
About this item
Systems for vacuum filtration of aqueous solutions including cell culture media, buffers, or other biological fluids.
- Polystyrene funnel and collection reservoir
- Each individual unit is lot numbered for traceability
- Solution can be stored in the collection reservoir until needed
- Top filter and bottom collector bottle have engraved graduation
- Designed hose connector can fit multiple hose diameters
PES/PVDF/Nylon/SFCA membrane: Intended for the sterile vacuum filtration of aqueous solutions such as cell culture media and biological fluids.
Ideal for tissue culture applications.
Certifications: ISO 10993-5, 1999. Non pyrogenic. non cytotoxic.
Ordering information: Includes the filtration funnel, vacuum port, lid, collection reservoir, and cap for storage.
Packaging: Individually packaged in easy to peel bags. Receiver bottle cap is individually wrapped.
Customers Who Bought This Also Bought