Snap Caps, with Single Thumb Tab, Globe Scientific
Catalog #: 10040-008
Supplier: Globe Scientific
Specifications
- Color:Yellow
- Material:LDPE
- Fits:13 mm tubes
- Supplier No.:113146Y
Specifications
About this item
These colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes, and plastic culture tubes
- Low density polyethylene (LDPE)
- Assorted colors available
- Fits both glass and plastic tubes
Each cap is precision molded from low-density polyethylene, and maintains a tight seal during centrifugation, agitation, and refrigeration procedures. They are available in bright attractive colors for sample identification purposes. The caps feature a single thumb tab for easy removal of the cap. This feature also allows the tubes to be stacked closer in racks when compared to double thumb tab caps.
Customers Who Bought This Also Bought