Corning® Spheroid Microplates, Corning
Catalog #: 10037-556
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Sterility:Sterile
- Color:Black/Clear
- Description:Spheroid Mircoplates
- Environmentally Preferable:Low Manufacturing Impact
- No. of wells:384
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Cat. no.:10037-556
- Working Volume:20 - 90 µl
- Surface Treatment:Ultra-low attachment
- Bottom Style:Round
- Supplier No.:3830
Specifications
About this item
PS, black opaque microplate with optically clear bottom, with lid. The Ultra-Low Attachment surface is a covalently bound hydrogel layer that is hydrophilic and neutrally charged. The surface minimizes cell attachment, protein absorption, and enzyme activation. It is ideal for culturing stem cells, as it promotes embryoid body formation from ES cells.
- Well geometry aids formation of spheroids in centre of well
- Non cytotoxic, biologically inert and non degradable surface
- Sterilized by gamma radiation
Customers Who Bought This Also Bought